missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Avidin, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against native Avidin.
Brand: Abnova PAB9917.100ug
This item is not returnable.
View return policy
Specifications
| Avidin | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against native Avidin. | |
| In 20mM potassium phosphate buffer, 150mM NaCl, pH 7.2 (0.01% sodium azide) | |
| Native purified Avidin from hen egg white. | |
| RUO | |
| Chicken | |
| Lyophilized |
| ELISA, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| ELISA (1:2000-1:20000) Western Blot (1:1000-1:10000) Immunohistochemistry (1:1000-1:5000) The optimal working dilution should be determined by the end user. | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Store at 4°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction