Learn More
AVPR1B Rabbit anti-Human, Monkey, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of AVPR1B.
Brand: Abnova PAB25517.50ug
Description
The protein encoded by this gene acts as receptor for arginine vasopressin. This receptor belongs to the subfamily of G-protein coupled receptors which includes AVPR1A, V2R and OXT receptors. Its activity is mediated by G proteins which stimulate a phosphatidylinositol-calcium second messenger system. The receptor is primarily located in the anterior pituitary, where it stimulates ACTH release. It is expressed at high levels in ACTH-secreting pituitary adenomas as well as in bronchial carcinoids responsible for the ectopic ACTH syndrome. A spliced antisense transcript of this gene has been reported but its function is not known. [provided by RefSeq
Specifications
| AVPR1B | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of AVPR1B. | |
| In PBS (0.09% sodium azide) | |
| AVPR3 | |
| Rabbit | |
| Immunoaffinity chromatography | |
| RUO | |
| 553 | |
| Human, Monkey | |
| Liquid |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (2-10 ug/mL) The optimal working dilution should be determined by the end user. | |
| AVPR1B | |
| AVPR1B | |
| A synthetic peptide corresponding to 16 amino acids at 3rd cytoplasmic domain of human AVPR1B. | |
| 50 μg | |
| Primary | |
| BLAST analysis of the peptide immunogen showed no homology with other human proteins. | |
| Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only