Learn More
BACE1 Rabbit anti-Human, Monkey, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of BACE1.
Brand: Abnova PAB16471.50ug
Description
Cerebral deposition of amyloid beta peptide is an early and critical feature of Alzheimer's disease. Amyloid beta peptide is generated by proteolytic cleavage of amyloid precursor protein (APP) by two proteases, one of which is the protein encoded by this gene. The encoded protein, a member of the peptidase A1 protein family, is a type I integral membrane glycoprotein and aspartic protease that is found mainly in the Golgi. Four transcript variants encoding different isoforms have been described for this gene. [provided by RefSeq]
Specifications
| BACE1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of BACE1. | |
| In PBS (0.09% sodium azide) | |
| P56817 | |
| BACE1 | |
| A synthetic peptide (conjugated with KLH) corresponding to human BACE1. | |
| 50 μg | |
| Primary | |
| Near the N-terminus of human. | |
| Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (5 ug/mL) The optimal working dilution should be determined by the end user. | |
| BACE1 | |
| ASP2/BACE/FLJ90568/HSPC104/KIAA1149 | |
| Rabbit | |
| Immunoaffinity purification | |
| RUO | |
| 23621 | |
| Human, Monkey | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only