missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BBC3 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of BBC3.
Brand: Abnova PAB13106.100ug
This item is not returnable.
View return policy
Specifications
| BBC3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of BBC3. | |
| In PBS (0.02% sodium azide) | |
| NP_055232 | |
| BBC3 | |
| A synthetic peptide corresponding to N-terminus 14 amino acids of human BBC3. | |
| RUO | |
| 27113 | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Western Blot (2-4 ug/mL) The optimal working dilution should be determined by the end user. | |
| BBC3 | |
| JFY1/PUMA | |
| Rabbit | |
| 100 μg | |
| Primary | |
| This sequence is identical between a and b forms of the PUMA proteins. K-562 cell lysate can be used as a positive control and a band at approximately 23 KDa can be detected. antibody can also detect PUMA by immunohistochemistry at 10 ug/mL. | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction