Learn More
BCL2 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of BCL2.
Brand: Abnova PAB13118.100ug
Description
This gene encodes an integral outer mitochondrial membrane protein that blocks the apoptotic death of some cells such as lymphocytes. Constitutive expression of BCL2, such as in the case of translocation of BCL2 to Ig heavy chain locus, is thought to be the cause of follicular lymphoma. Two transcript variants, produced by alternate splicing, differ in their C-terminal ends. [provided by RefSeq
Specifications
| BCL2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of BCL2. | |
| In PBS (0.02% sodium azide) | |
| AAH27258 | |
| BCL2 | |
| A synthetic peptide corresponding to N-terminus 15 amino acids of human BCL2. | |
| RUO | |
| 596 | |
| Human, Mouse | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Western Blot (1-2 ug/mL) Immunohistochemistry (2 ug/mL) The optimal working dilution should be determined by the end user. | |
| BCL2 | |
| Bcl-2 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| It is human and mouse reactive. | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only