Learn More
BCL6 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of BCL6.
Brand: Abnova PAB25644.100ug
Description
The protein encoded by this gene is a zinc finger transcription factor and contains an N-terminal POZ domain. This protein acts as a sequence-specific repressor of transcription, and has been shown to modulate the transcription of START-dependent IL-4 responses of B cells. This protein can interact with a variety of POZ-containing proteins that function as transcription corepressors. This gene is found to be frequently translocated and hypermutated in diffuse large-cell lymphoma (DLCL), and may be involved in the pathogenesis of DLCL. Alternatively spliced transcript variants encoding different protein isoforms have been found for this gene. [provided by RefSeq
Specifications
| BCL6 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user. | |
| BCL6 | |
| BCL5/BCL6A/LAZ3/ZBTB27/ZNF51 | |
| Rabbit | |
| Peptide affinity purification | |
| RUO | |
| 604 | |
| Human, Mouse, Rat | |
| Liquid |
| ELISA, Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of BCL6. | |
| In PBS (0.05% sodium azide) | |
| NP_001124317 | |
| BCL6 | |
| A synthetic peptide corresponding to 18 amino acids at internal region of human BCL6. | |
| 100 μg | |
| Primary | |
| Three alternatively spliced transcript variants have been observed. | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only