Learn More
BDNF Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of BDNF.
Brand: Abnova PAB7956.100ug
Description
The protein encoded by this gene is a member of the nerve growth factor family. It is induced by cortical neurons, and is necessary for survival of striatal neurons in the brain. Expression of this gene is reduced in both Alzheimer's and Huntington disease patients. This gene may play a role in the regulation of stress response and in the biology of mood disorders. Multiple transcript variants encoding distinct isoforms have been described for this gene. [provided by RefSeq]
Specifications
| BDNF | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of BDNF. | |
| Lyophilized from 0.9mg NaCl, 0.2mg Na2HPO4 (5mg BSA, 0.05mg sodium azide, 0.05mg Thimerosal) | |
| MGC34632 | |
| Rabbit | |
| Immunoaffinity purification | |
| RUO | |
| 627 | |
| Human, Mouse, Rat | |
| Lyophilized |
| Western Blot | |
| Unconjugated | |
| Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user. | |
| BDNF | |
| BDNF | |
| A synthetic peptide corresponding to amino acids at middle region of human BDNF. | |
| 100 μg | |
| Primary | |
| Identical to the related rat and mouse sequence. | |
| Store at -20°C on dry atmosphere. After reconstitution with 200μL of deionized water and concentration will be 500μ/mL store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only