missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BECN1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of BECN1.
Brand: Abnova PAB0728.100uL
This item is not returnable.
View return policy
Specifications
| BECN1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of BECN1. | |
| Lyophilized from PBS | |
| Q14457 | |
| BECN1 | |
| A synthetic peptide (conjugated with Blue Carrier Protein) corresponding to C-terminus of human BECN1. | |
| RUO | |
| 8678 | |
| Human | |
| Lyophilized |
| Immunofluorescence, Immunohistochemistry (Frozen), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:100-1:2000) Immunofluorescence (1:100-1:2000) Western Blot (1:100-1:2000) The optimal working dilution should be determined by the end user. | |
| BECN1 | |
| ATG6/VPS30/beclin1 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Specific for Beclin-1. | |
| Store at 4°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction