missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beta-Galactosidase Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against Beta-Galactosidase.
Brand: Abnova PAB13740.100ug
This item is not returnable.
View return policy
Specifications
| Beta-Galactosidase | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against Beta-Galactosidase. | |
| In PBS, pH 7.2 (0.09% sodium azide) | |
| Native Escherichia coli Beta-Galactosidase. | |
| RUO | |
| E. coli | |
| Liquid |
| ELISA, Western Blot | |
| Unconjugated | |
| Western Blot (1:1000-1:3000) The optimal working dilution should be determined by the end user. | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Store at 4°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction