missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BGLAP, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of BGLAP.
Brand: Abnova PAB3924.400uL
This item is not returnable.
View return policy
Specifications
| BGLAP | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of BGLAP. | |
| In PBS (0.09% sodium azide) | |
| NP_000702 | |
| BGLAP | |
| A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human BGLAP. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:50-100) Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| BGLAP | |
| BGP/OC/PMF1 | |
| Rabbit | |
| Protein G purification | |
| RUO | |
| 632 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction