Learn More
BIRC5, Rabbit, DyLight 488, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of BIRC5.
Brand: Abnova PAB15181.100uL
Description
This gene is a member of the inhibitor of apoptosis (IAP) gene family, which encode negative regulatory proteins that prevent apoptotic cell death. IAP family members usually contain multiple baculovirus IAP repeat (BIR) domains, but this gene encodes proteins with only a single BIR domain. The encoded proteins also lack a C-terminus RING finger domain. Gene expression is high during fetal development and in most tumors yet low in adult tissues. Antisense transcripts are involved in the regulation of this gene's expression. At least four transcript variants encoding distinct isoforms have been found for this gene, but the full-length natures of only three of them have been determined. [provided by RefSeq]
Specifications
| BIRC5 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic phosphopeptide of BIRC5. | |
| In 50mM sodium borate | |
| O15392 | |
| BIRC5 | |
| Synthetic phosphopeptide corresponding to residues surrounding S20 of human BIRC5. | |
| RUO | |
| 332 | |
| Human | |
| Liquid |
| Western Blot | |
| DyLight 488 | |
| Western Blot (3 ug/mL) The optimal working dilution should be determined by the end user. | |
| BIRC5 | |
| API4/EPR-1 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| This antibody is useful for Western blot, where a band is seen ∼16.5 KDa. | |
| Store at 4°C. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only