missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BMP8A Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of BMP8A.
Brand: Abnova PAB25100.100uL
This item is not returnable.
View return policy
Specifications
| BMP8A | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:200) Immunofluorescence (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| BMP8A | |
| BMP8A | |
| A synthetic peptide corresponding to BMP8A. | |
| 100 μL | |
| Primary | |
| BMP8A polyclonal antibody detects endogenous levels of BMP8A protein. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of BMP8A. | |
| In PBS, pH 7.2 (0.05% sodium azide) | |
| FLJ14351/FLJ45264 | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 353500 | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction