missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Bovine IgG whole molecule,Polyclonal Secondary Antibody, Abnova™
Brand: Abnova PAB9148.2mL
This item is not returnable.
View return policy
Specifications
| Rabbit anti-Bovine IgG whole molecule | |
| Polyclonal | |
| Rabbit anti-bovine IgG recognizes bovine IgG whole molecule. This secondary antibody was purified using delipidation and defibrination. | |
| Lyophilized from 10mM KH2PO4, 150mM NaCl, pH 7.2 | |
| Bovine IgG whole molecule | |
| 2 mL | |
| Bovine | |
| Lyophilized |
| ELISA, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| Rabbit | |
| This product was prepared from monospecific antiserum by a delipidation and defibrination. Assay by immunoelectrophoresis resulted in a single precipitin arc against bovine IgG and bovine serum. | |
| Secondary | |
| Store at 4°C on dry atmosphere. After reconstitution with 2.0mL of deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction