Learn More
BUB1B Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of BUB1B.
Brand: Abnova PAB26883.100uL
Description
This gene encodes a kinase involved in spindle checkpoint function. The protein has been localized to the kinetochore and plays a role in the inhibition of the anaphase-promoting complex/cyclosome (APC/C), delaying the onset of anaphase and ensuring proper chromosome segregation. Impaired spindle checkpoint function has been found in many forms of cancer. [provided by RefSeq
Specifications
| BUB1B | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user. | |
| BUB1B | |
| BUB1beta/BUBR1/Bub1A/MAD3L/SSK1/hBUBR1 | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 701 | |
| Human, Mouse | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of BUB1B. | |
| In PBS, pH 7.2 (0.05% sodium azide) | |
| O60566 | |
| BUB1B | |
| A synthetic peptide corresponding to human BUB1B. | |
| 100 μL | |
| Primary | |
| BUB1B polyclonal antibody detects endogenous levels of BUB1B protein. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only