missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C22orf28, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant C22orf28.
Brand: Abnova PAB20033.100uL
This item is not returnable.
View return policy
Description
Sequence: PDDLDLHVIYDVSHNIAKVEQHVVDGKERTLLVHRKGSTRAFPPHHPLIAVDYQLTGQPVLIGGTMGTCSYVLTGTEQGMTETFGTTCHGAGRALSRAKSRRNLDFQDVLDKLADMGIAIRVASPKLVMEEAPESYKNVTDVVNTCHDAGISKKAIKLRPSpecifications
| C22orf28 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant C22orf28. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| Q9Y3I0 | |
| C22orf28 | |
| Recombinant protein corresponding to amino acids of human C22orf28. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:20-1:50) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
| C22orf28 | |
| DJ149A16.6/HSPC117/RP1-149A16.6 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 51493 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Product Content Correction
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
For Research Use Only
Ser du en mulighed for forbedring?Del en indholdskorrektion