Learn More
C8B, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant C8B.
Brand: Abnova PAB21600.100uL
Description
C8 beta is one of the three subunits that comprise the component 8 (C8) of the complement system. C8 participates in the formation of Membrane Attack Complex that results in the lysis of cells. Patients with C8B deficiency are prone to bacteria infection. [provided by RefSeq
Sequence: PCQGNGVPVLKGSRCDCICPVGSQGLACEVSYRKNTPIDGKWNCWSNWSSCSGRRKTRQRQCNNPPPQNGGSPCSGPASETLDCSSpecifications
| C8B | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant C8B. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| P07358 | |
| C8B | |
| Recombinant protein corresponding to amino acids of human C8B. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:10-1:20) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
| C8B | |
| MGC163447 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 732 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only