missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cacna1c, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of Cacna1c.
Brand: Abnova PAB9795.100ug
This item is not returnable.
View return policy
Specifications
| Cacna1c | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of Cacna1c. | |
| In PBS (0.08% sodium azide) | |
| RATIVS302 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Liquid |
| Western Blot | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| Cacna1c | |
| Cacna1c | |
| A synthetic peptide (conjugated with KLH) corresponding to rat Cacna1c. | |
| RUO | |
| 24239 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction