Learn More
CARD10 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CARD10.
Brand: Abnova PAB12878.100ug
Description
The caspase recruitment domain (CARD) is a protein module that consists of 6 or 7 antiparallel alpha helices. It participates in apoptosis signaling through highly specific protein-protein homophilic interactions. Like several other CARD proteins, CARD10 belongs to the membrane-associated guanylate kinase (MAGUK) family and activates NF-kappa-B (NFKB; see MIM 164011) through BCL10 (MIM 603517) (Wang et al., 2001 [PubMed 11259443]).[supplied by OMIM
Specifications
| CARD10 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of CARD10. | |
| In PBS (0.02% sodium azide) | |
| AAK26165 | |
| CARD10 | |
| A synthetic peptide corresponding to C-terminus 14 amino acids of human CARD10. | |
| RUO | |
| 29775 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Immunocytochemistry, Western Blot | |
| Unconjugated | |
| Western Blot (5-10 ug/mL) The optimal working dilution should be determined by the end user. | |
| CARD10 | |
| BIMP1/CARMA3/MGC142219 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only