Learn More
CASP9 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CASP9.
Brand: Abnova PAB24855.100uL
Description
This gene encodes a member of the cysteine-aspartic acid protease (caspase) family. Sequential activation of caspases plays a central role in the execution-phase of cell apoptosis. Caspases exist as inactive proenzymes which undergo proteolytic processing at conserved aspartic residues to produce two subunits, large and small, that dimerize to form the active enzyme. This protein is processed by caspase APAF1; this step is thought to be one of the earliest in the caspase activation cascade. Alternative splicing results in two transcript variants which encode different isoforms. [provided by RefSeq
Specifications
| CASP9 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of CASP9. | |
| In 1X PBS, pH 7.2 (0.09% sodium azide) | |
| APAF-3/APAF3/CASPASE-9c/ICE-LAP6/MCH6 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 842 | |
| Human | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:200) Immunofluorescence (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| CASP9 | |
| CASP9 | |
| A synthetic peptide corresponding to CASP9. | |
| 100 μL | |
| Primary | |
| CASP9 polyclonal antibody detects endogenous levels of CASP9 protein. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only