Learn More
CBLC Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CBLC.
Brand:  Abnova PAB1698.400uL
Product Code. 16022527
  • € 388.00 / 400µL
Description
CBL proteins, such as CBLC, are phosphorylated upon activation of a variety of receptors that signal via protein tyrosine kinases. Through interactions with proteins containing SRC (MIM 190090) homology-2 (SH2) and SH3 domains, CBL proteins modulate downstream cell signaling (Keane et al., 1999 [PubMed 10362357]).[supplied by OMIM
Specifications
Specifications
CBLC
Polyclonal
Rabbit polyclonal antibody raised against synthetic peptide of CBLC.
In PBS (0.09% sodium azide)
NP_036248;Q9ULV8
CBLC
A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human CBLC.
400 μL
Primary
Human
Liquid
ELISA, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Western Blot (1:1000) Immunohistochemistry (1:50-100) The optimal working dilution should be determined by the end user.
CBLC
CBL-3/CBL-SL/RNF57
Rabbit
Protein G purification
RUO
23624
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.