missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCDC22 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CCDC22.
Brand: Abnova PAB27201.100ug
This item is not returnable.
View return policy
Specifications
| CCDC22 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user. | |
| CCDC22 | |
| CXorf37/JM1 | |
| Rabbit | |
| Peptide affinity purification | |
| RUO | |
| 28952 | |
| Human, Mouse, Rat | |
| Liquid |
| ELISA, Immunofluorescence, Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of CCDC22. | |
| In PBS (0.02% sodium azide) | |
| NP_055588 | |
| CCDC22 | |
| A synthetic peptide corresponding to 18 amino acids at N-terminus of human CCDC22. | |
| 100 μg | |
| Primary | |
| At least three isoforms of CCDC22 are known to exist; this antibody will detect the two largest isoforms. | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction