missing translation for 'onlineSavingsMsg'
Learn More

CCDC88A Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™

Product Code. 16042757
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16042757 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16042757 Supplier Abnova Supplier No. PAB17324.100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit polyclonal antibody raised against synthetic peptide of CCDC88A.

Specifications

Antigen CCDC88A
Applications Immunohistochemistry (PFA fixed)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of CCDC88A.
Dilution Immunohistochemistry (1:50-1:100) ELISA (1:5000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol)
Gene CCDC88A
Gene Accession No. Q3V6T2
Gene Alias APE/DKFZp686D0630/GIRDIN/GIV/GRDN/HkRP1/KIAA1212
Gene Symbols CCDC88A
Host Species Rabbit
Immunogen A synthetic peptide corresponding to internal of human CCDC88A.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55704
Test Specificity This antibody detects endogenous levels of total CCDC88A protein.
Target Species Human, Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.