Learn More
CCNB1 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against full-length recombinant CCNB1.
Brand: Abnova PAB9943.100uL
Description
The protein encoded by this gene is a regulatory protein involved in mitosis. The gene product complexes with p34(cdc2) to form the maturation-promoting factor (MPF). Two alternative transcripts have been found, a constitutively expressed transcript and a cell cycle-regulated transcript, that is expressed predominantly during G2/M phase. The different transcripts result from the use of alternate transcription initiation sites. [provided by RefSeq
Specifications
| CCNB1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against full length recombinant CCNB1. | |
| In antiserum (0.01% sodium azide) | |
| NP_114172;P14635 | |
| CCNB1 | |
| Recombinant GST fusion protein corresponding to full length human CCNB1. | |
| RUO | |
| 891 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot | |
| Unconjugated | |
| ELISA (1:2000-1:10000) Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user. | |
| CCNB1 | |
| CCNB | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only