missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCNO Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CCNO.
Brand: Abnova PAB13206.100ug
This item is not returnable.
View return policy
Specifications
| CCNO | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user. | |
| CCNO | |
| CCNU/FLJ22422/UDG2/UNG2 | |
| Rabbit | |
| Peptide affinity purification | |
| RUO | |
| 10309 | |
| Human, Mouse, Rat | |
| Liquid |
| ELISA, Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of CCNO. | |
| In PBS (0.02% sodium azide) | |
| NP_066970 | |
| CCNO | |
| A synthetic peptide corresponding to 13 amino acids near internal region of human CCNO. | |
| 100 μg | |
| Primary | |
| At least two isoforms of Cyclin O are known to exist; this antibody will recognize both isoforms. | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction