Learn More
CCR6 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CCR6.
Brand: Abnova PAB8939.100ug
Description
This gene encodes a member of the beta chemokine receptor family, which is predicted to be a seven transmembrane protein similar to G protein-coupled receptors. The gene is preferentially expressed by immature dendritic cells and memory T cells. The ligand of this receptor is macrophage inflammatory protein 3 alpha (MIP-3 alpha). This receptor has been shown to be important for B-lineage maturation and antigen-driven B-cell differentiation, and it may regulate the migration and recruitment of dentritic and T cells during inflammatory and immunological responses. Alternatively spliced transcript variants that encode the same protein have been described for this gene. [provided by RefSeq
Sequence: DYFVSVNTSYYSVDSEMLLCSpecifications
| CCR6 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of CCR6. | |
| In buffer containing 0.02% sodium azide | |
| BN-1/CD196/CKR-L3/CKR6/CKRL3/CMKBR6/DCR2/DRY-6/GPR-CY4/GPR29/GPRCY4/STRL22 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Western Blot (1:500) Immunoprecipitation (1:250) The optimal working dilution should be determined by the end user. | |
| CCR6 | |
| CCR6 | |
| A synthetic peptide corresponding to amino acids 17-36 of human CCR6. | |
| RUO | |
| 1235 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only