Learn More
CD180 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CD180.
Brand: Abnova PAB13431.100ug
Description
CD180 is a cell surface molecule consisting of extracellular leucine-rich repeats (LRR) and a short cytoplasmic tail. The extracellular LRR is associated with a molecule called MD-1 and form the cell surface receptor complex, RP105/MD-1. It belongs to the family of pathogen receptors, Toll-like receptors (TLR). RP105/MD1, by working in concert with TLR4, controls B cell recognition and signaling of lipopolysaccharide (LPS), a membrane constituent of Gram-negative bacteria. [provided by RefSeq
Specifications
| CD180 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of CD180. | |
| In PBS (0.02% sodium azide) | |
| BAA12019 | |
| CD180 | |
| A synthetic peptide corresponding to C-terminus 14 amino acids of human CD180. | |
| RUO | |
| 4064 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Western Blot (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| CD180 | |
| LY64/Ly78/MGC126233/MGC126234/RP105 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only