missing translation for 'onlineSavingsMsg'
Learn More

Cd97, Rabbit, Polyclonal Antibody, Abnova™

Product Code. 16202780
Change view
Click to view available options
Quantity:
500 μg
Unit Size:
500µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16202780 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16202780 Supplier Abnova Supplier No. PAB10186.500ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit polyclonal antibody raised against partial recombinant Cd97.

Specifications

Antigen Cd97
Applications ELISA, Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against partial recombinant Cd97.
Dilution Western Blot (1:1000-1:5000) Immunofluorescence (1:1000-1:5000) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from 20mM potassium phosphate buffer, 150mM NaCl, pH 7.2 (0.01% sodium azide)
Gene Cd97
Gene Accession No. NP_036055;Q9Z0M6
Gene Alias AA409984/TM7LN1
Gene Symbols Cd97
Host Species Rabbit
Immunogen Recombinant protein corresponding to amino acids 1-512 of mouse Cd97.
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26364
Target Species Mouse, Rat
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot after reconstitution to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.