Learn More
CDC25C Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CDC25C.
Brand: Abnova PAB24805.100uL
Description
This gene is highly conserved during evolution and it plays a key role in the regulation of cell division. The encoded protein is a tyrosine phosphatase and belongs to the Cdc25 phosphatase family. It directs dephosphorylation of cyclin B-bound CDC2 and triggers entry into mitosis. It is also thought to suppress p53-induced growth arrest. Multiple alternatively spliced transcript variants of this gene have been described, however, the full-length nature of many of them is not known. [provided by RefSeq
Specifications
| CDC25C | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| CDC25C | |
| CDC25C | |
| A synthetic peptide corresponding to CDC25C. | |
| 100 μL | |
| Primary | |
| CDC25C polyclonal antibody detects endogenous levels of CDC25C protein. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Immunohistochemistry (PFA fixed), Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of CDC25C. | |
| In PBS, pH 7.2 (0.05% sodium azide) | |
| CDC25 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 995 | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only