Learn More
CDK1 (phospho Y15), Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of CDK1.
Brand: Abnova PAB25833.100ug
Description
The protein encoded by this gene is a member of the Ser/Thr protein kinase family. This protein is a catalytic subunit of the highly conserved protein kinase complex known as M-phase promoting factor (MPF), which is essential for G1/S and G2/M phase transitions of eukaryotic cell cycle. Mitotic cyclins stably associate with this protein and function as regulatory subunits. The kinase activity of this protein is controlled by cyclin accumulation and destruction through the cell cycle. The phosphorylation and dephosphorylation of this protein also play important regulatory roles in cell cycle control. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: G-T-Yp-G-VSpecifications
| CDK1 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user. | |
| CDC2 | |
| CDC28A/CDK1/DKFZp686L20222/MGC111195 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 983 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic phosphopeptide of CDK1. | |
| In PBS (without mg2+ and Ca2+), 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide) | |
| P06493 | |
| CDC2 | |
| Synthetic phosphopeptide corresponding to residues surrounding Y15 of human CDK1. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only