Learn More
CDKN1A Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CDKN1A.
Brand: Abnova PAB2922.400uL
Description
This gene encodes a potent cyclin-dependent kinase inhibitor. The encoded protein binds to and inhibits the activity of cyclin-CDK2 or -CDK4 complexes, and thus functions as a regulator of cell cycle progression at G1. The expression of this gene is tightly controlled by the tumor suppressor protein p53, through which this protein mediates the p53-dependent cell cycle G1 phase arrest in response to a variety of stress stimuli. This protein can interact with proliferating cell nuclear antigen (PCNA), a DNA polymerase accessory factor, and plays a regulatory role in S phase DNA replication and DNA damage repair. This protein was reported to be specifically cleaved by CASP3-like caspases, which thus leads to a dramatic activation of CDK2, and may be instrumental in the execution of apoptosis following caspase activation. Two alternatively spliced variants, which encode an identical protein, have been reported. [provided by RefSeq
Specifications
| CDKN1A | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of CDKN1A. | |
| In PBS (0.09% sodium azide) | |
| P38936 | |
| CDKN1A | |
| A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human CDKN1A. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Western Blot (1:1000) Immunohistochemistry (1:50-100) The optimal working dilution should be determined by the end user. | |
| CDKN1A | |
| CAP20/CDKN1/CIP1/MDA-6/P21/SDI1/WAF1/p21CIP1 | |
| Rabbit | |
| Protein G purification | |
| RUO | |
| 1026 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only