Learn More
CDKN1B Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against full-length recombinant CDKN1B.
Brand: Abnova PAB10300.100uL
Description
This gene encodes a cyclin-dependent kinase inhibitor, which shares a limited similarity with CDK inhibitor CDKN1A/p21. The encoded protein binds to and prevents the activation of cyclin E-CDK2 or cyclin D-CDK4 complexes, and thus controls the cell cycle progression at G1. The degradation of this protein, which is triggered by its CDK dependent phosphorylation and subsequent ubiquitination by SCF complexes, is required for the cellular transition from quiescence to the proliferative state. [provided by RefSeq
Specifications
| CDKN1B | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against full length recombinant CDKN1B. | |
| In 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide) | |
| P46527;NP_004055 | |
| CDKN1B | |
| Recombinant protein corresponding to full length human CDKN1B. | |
| RUO | |
| 1027 | |
| Human | |
| Liquid |
| ELISA, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| ELISA (1:10000-1:40000) Western Blot (1:1000-1:4000) The optimal working dilution should be determined by the end user. | |
| CDKN1B | |
| CDKN4/KIP1/MEN1B/MEN4/P27KIP1 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| Antiserum will specifically react with a p27 protein from human tissue. No reaction was observed against other related tumor suppressor proteins. No reactivity is observed with p27 from mouse. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only