Learn More
CDKN2A Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CDKN2A.
Brand: Abnova PAB12461.100uL
Description
This gene generates several transcript variants which differ in their first exons. At least three alternatively spliced variants encoding distinct proteins have been reported, two of which encode structurally related isoforms known to function as inhibitors of CDK4 kinase. The remaining transcript includes an alternate first exon located 20kb upstream of the remainder of the gene; this transcript contains an alternate open reading frame (ARF) that specifies a protein which is structurally unrelated to the products of the other variants. This ARF product functions as a stabilizer of the tumor suppressor protein p53 as it can interact with, and sequester, MDM1, a protein responsible for the degradation of p53. In spite of the structural and functional differences, the CDK inhibitor isoforms and the ARF product encoded by this gene, through the regulatory roles of CDK4 and p53 in cell cycle G1 progression, share a common functionality in cell cycle G1 control. This gene is frequently mutated or deleted in a wide variety of tumors, and is known to be an important tumor suppressor gene. [provided by RefSeq
Specifications
| CDKN2A | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of CDKN2A. | |
| In Tris-glycine, 150mM NaCl (0.05% sodium azide) | |
| Q8N726 | |
| CDKN2A | |
| A synthetic peptide corresponding to amino acids 100-200 of human CDKN2A. | |
| RUO | |
| 1029 | |
| Human | |
| Liquid |
| Western Blot | |
| Unconjugated | |
| Western Blot (2 ug/mL) The optimal working dilution should be determined by the end user. | |
| CDKN2A | |
| ARF/CDK4I/CDKN2/CMM2/INK4/INK4a/MLM/MTS1/TP16/p14/p14ARF/p16/p16INK4/p16INK4a/p19 | |
| Rabbit | |
| 100 μL | |
| Primary | |
| This antibody is specific to p14ARF. | |
| Store at -20°C or -80°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only