missing translation for 'onlineSavingsMsg'
Learn More

CDR2, Rabbit, Polyclonal Antibody, Abnova™

Product Code. 16099846
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16099846 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16099846 Supplier Abnova Supplier No. PAB12460.100uL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit polyclonal antibody raised against synthetic peptide of CDR2.

Specifications

Antigen CDR2
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of CDR2.
Dilution Western Blot (2 ug/mL) Immunohistochemistry (5-10 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In Tris-citrate/phosphate buffer, pH 7.0-8.0 (0.09% sodium azide)
Gene CDR2
Gene Accession No. Q01850
Gene Alias CDR62/Yo
Gene Symbols CDR2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to C-terminus of human CDR2.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1039
Target Species Bovine, Human, Mouse, Primate
Content And Storage Store at 4°C. Do not freeze.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.