Learn More
CEBPB Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CEBPB.
Brand: Abnova PAB27156.100uL
Description
The protein encoded by this intronless gene is a bZIP transcription factor which can bind as a homodimer to certain DNA regulatory regions. It can also form heterodimers with the related proteins CEBP-alpha, CEBP-delta, and CEBP-gamma. The encoded protein is important in the regulation of genes involved in immune and inflammatory responses and has been shown to bind to the IL-1 response element in the IL-6 gene, as well as to regulatory regions of several acute-phase and cytokine genes. In addition, the encoded protein can bind the promoter and upstream element and stimulate the expression of the collagen type I gene. [provided by RefSeq
Specifications
| CEBPB | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of CEBPB. | |
| In PBS, pH 7.2 (0.09% sodium azide) | |
| P17676 | |
| CEBPB | |
| A synthetic peptide corresponding to human CEBPB. | |
| 100 μL | |
| Primary | |
| CEBPB polyclonal antibody detects endogenous levels of CEBPB protein. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| CEBPB | |
| C/EBP-beta/CRP2/IL6DBP/LAP/MGC32080/NF-IL6/TCF5 | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 1051 | |
| Human, Mouse | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only