Learn More
CELSR3 Rabbit anti-Bovine, Hamster, Horse, Human, Monkey, Mouse, Rabbit, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CELSR3.
Brand: Abnova PAB26117.50ug
Description
The protein encoded by this gene is a member of the flamingo subfamily, part of the cadherin superfamily. The flamingo subfamily consists of nonclassic-type cadherins; a subpopulation that does not interact with catenins. The flamingo cadherins are located at the plasma membrane and have nine cadherin domains, seven epidermal growth factor-like repeats and two laminin A G-type repeats in their ectodomain. They also have seven transmembrane domains, a characteristic unique to this subfamily. It is postulated that these proteins are receptors involved in contact-mediated communication, with cadherin domains acting as homophilic binding regions and the EGF-like domains involved in cell adhesion and receptor-ligand interactions. The specific function of this particular member has not been determined. [provided by RefSeq
Specifications
| CELSR3 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of CELSR3. | |
| In PBS (0.09% sodium azide) | |
| Q9NYQ7 | |
| CELSR3 | |
| A synthetic peptide corresponding to 15 amino acids at N-terminus of human CELSR3. | |
| 50 μg | |
| Primary | |
| BLAST analysis of the peptide immunogen showed no homology with other human proteins. | |
| Store at 4°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. |
| Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (4-11 ug/mL) The optimal working dilution should be determined by the end user. | |
| CELSR3 | |
| CDHF11/EGFL1/FMI1/HFMI1/MEGF2/RESDA1 | |
| Rabbit | |
| Immunoaffinity chromatography | |
| RUO | |
| 1951 | |
| Bovine, Equine, Hamster, Human, Monkey, Mouse, Rabbit, Rat | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only