missing translation for 'onlineSavingsMsg'
Learn More

CESA1 (phospho T227), Rabbit anti-Thale cress, Polyclonal Antibody, Abnova™

Product Code. 16152090
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16152090 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16152090 Supplier Abnova Supplier No. PAB15902.100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit polyclonal antibody raised against synthetic phosphopeptide of CESA1.

Specifications

Antigen CESA1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic phosphopeptide of CESA1.
Dilution ELISA (1:2000-1:5000) Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (50% glycerol, 0.01% sodium azide)
Gene CESA1
Gene Alias CELLULOSE SYNTHASE 1/F8B4.110/F8B4_110/RADIALLY SWOLLEN 1/RSW1
Gene Symbols CESA1
Host Species Rabbit
Immunogen Synthetic phosphopeptide corresponding to residues surrounding T227 of Arabidopsis thaliana CESA1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 829376
Target Species Thale cress
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.