Learn More
CFTR Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CFTR.
Brand: Abnova PAB18325.100ug
Description
This gene encodes a member of the ATP-binding cassette (ABC) transporter superfamily. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the MRP subfamily that is involved in multi-drug resistance. The encoded protein functions as a chloride channel and controls the regulation of other transport pathways. Mutations in this gene are associated with the autosomal recessive disorders cystic fibrosis and congenital bilateral aplasia of the vas deferens. Alternatively spliced transcript variants have been described, many of which result from mutations in this gene. [provided by RefSeq
Specifications
| CFTR | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:100) ELISA (1:40000) The optimal working dilution should be determined by the end user. | |
| CFTR | |
| ABC35/ABCC7/CF/CFTR/MRP/MRP7/TNR-CFTR/dJ760C5.1 | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 1080 | |
| Human, Mouse, Rat | |
| Liquid |
| ELISA, Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of CFTR. | |
| In PBS, 150mM NaCl, pH 7.4 (50% glycerol, 0.02% sodium azide) | |
| P13569 | |
| CFTR | |
| A synthetic peptide corresponding to residues surrounding S737 of human CFTR. | |
| 100 μg | |
| Primary | |
| This antibody is specific to CFTR. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only