missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rabbit anti-Chicken IgG Fc Secondary Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-62593
This item is not returnable.
View return policy
Description
IgG Fc Polyclonal antibody specifically detects IgG Fc in Chicken samples. It is validated for Western Blot, ELISA, Immunohistochemistry
Specifications
| IgG Fc | |
| Polyclonal | |
| Western Blot 1:2000-1:10000, ELISA 1:20000 - 1:100000, Immunohistochemistry 1:1000 - 1:5000 | |
| Rabbit | |
| Multi-step | |
| RUO | |
| Secondary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry | |
| Unconjugated | |
| 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 | |
| Chicken IgG F(c) fragment | |
| 2 mg | |
| Adaptive Immunity, Immunology | |
| Chicken | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction