Learn More
CHKA Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CHKA.
Brand: Abnova PAB4068.400uL
Description
The major pathway for the biosynthesis of phosphatidylcholine occurs via the CDP-choline pathway. The protein encoded by this gene is the initial enzyme in the sequence and may play a regulatory role. The encoded protein also catalyzes the phosphorylation of ethanolamine. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Specifications
| CHKA | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of CHKA. | |
| In PBS (0.09% sodium azide) | |
| NP_997634;NP_001268 | |
| CHKA | |
| A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human CHKA. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| Unconjugated | |
| ELISA (1:1000) Western Blot (1:100-500) The optimal working dilution should be determined by the end user. | |
| CHKA | |
| CHK/CKI | |
| Rabbit | |
| Protein G purification | |
| RUO | |
| 1119 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only