missing translation for 'onlineSavingsMsg'
Learn More

Cideb, Rabbit, Polyclonal Antibody, Abnova™

Product Code. 16009576
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16009576 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16009576 Supplier Abnova Supplier No. PAB11961.100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit polyclonal antibody raised against synthetic peptide of Cideb.

Specifications

Antigen Cideb
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of Cideb.
Dilution Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (0.02% sodium azide)
Gene Cideb
Gene Accession No. AF041377
Gene Alias 1110030C18Rik/AI790179/CIDE-B
Gene Symbols Cideb
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 204-219 of mouse Cideb.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12684
Test Specificity This antibody recognizes mouse CIDE-B (25kD). No cross-reactivity with CIDE-A. Epitopes at the C-terminus of CIDE-B.
Target Species Mouse
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.