Learn More
CLIC3, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CLIC3.
Brand: Abnova PAB25197.100uL
Description
Chloride channels are a diverse group of proteins that regulate fundamental cellular processes including stabilization of cell membrane potential, transepithelial transport, maintenance of intracellular pH, and regulation of cell volume. Chloride intracellular channel 3 is a member of the p64 family and is predominantly localized in the nucleus and stimulates chloride ion channel activity. In addition, this protein may participate in cellular growth control, based on its association with ERK7, a member of the MAP kinase family. [provided by RefSeq
Specifications
| CLIC3 | |
| Polyclonal | |
| Unconjugated | |
| Western Blot (1:500-1:1000) Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
| CLIC3 | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| 9022 | |
| Human | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| Rabbit polyclonal antibody raised against synthetic peptide of CLIC3. | |
| In PBS, pH 7.2 (0.05% sodium azide) | |
| CLIC3 | |
| A synthetic peptide corresponding to CLIC3. | |
| 100 μL | |
| Primary | |
| CLIC3 polyclonal antibody detects endogenous levels of CLIC3 protein. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only