missing translation for 'onlineSavingsMsg'
Learn More

CLIP1 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™

Product Code. 16027177
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16027177 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16027177 Supplier Abnova Supplier No. PAB26994.100uL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit polyclonal antibody raised against synthetic peptide of CLIP1.

Specifications

Antigen CLIP1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptide of CLIP1.
Dilution Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (0.05% sodium azide)
Gene CLIP1
Gene Accession No. P30622
Gene Alias CLIP/CLIP-170/CLIP170/CYLN1/MGC131604/RSN
Gene Symbols CLIP1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to human CLIP1.
Purification Method Antigen affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6249
Test Specificity CLIP1 polyclonal antibody detects endogenous levels of CLIP1 protein.
Target Species Human, Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.