Learn More
CLN5, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant CLN5.
Brand: Abnova PAB27910.100uL
Description
This gene is one of eight which have been associated with neuronal ceroid lipofuscinoses (NCL). Also referred to as Batten disease, NCL comprises a class of autosomal recessive, neurodegenerative disorders affecting children. The genes responsible likely encode proteins involved in the degradation of post-translationally modified proteins in lysosomes. The primary defect in NCL disorders is thought to be associated with lysosomal storage function
Sequence: TLTGKNYTMEWYELFQLGNCTFPHLRPEMDAPFWCNQGAACFFEGIDDVHWKENGTLVQVATISGNMFNQMAKWVKQDNETGIYYETWNVKASPEKSpecifications
| CLN5 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant CLN5. | |
| In PBS, pH 7.2, (40% glycerol, 0.02% sodium azide) | |
| FLJ90628/NCL | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 1203 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| Immunohistochemistry (PFA fixed), Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:500-1:1000) Western Blot (1:100-1:250) The optimal working dilution should be determined by the end user. | |
| CLN5 | |
| CLN5 | |
| Recombinant protein corresponding to amino acids of human CLN5. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only