missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CNTD2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CNTD2.
Brand: Abnova PAB17510.100ug
This item is not returnable.
View return policy
Specifications
| CNTD2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of CNTD2. | |
| In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol) | |
| Q9H8S5 | |
| CNTD2 | |
| A synthetic peptide corresponding to internal of human CNTD2. | |
| RUO | |
| 79935 | |
| Human | |
| Liquid |
| ELISA, Immunohistochemistry (PFA fixed) | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:100) ELISA (1:40000) The optimal working dilution should be determined by the end user. | |
| CNTD2 | |
| FLJ13265 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| This antibody detects endogenous levels of total CNTD2 protein. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction