missing translation for 'onlineSavingsMsg'
Learn More

Col1a1/Col1a2 Rabbit anti-Rat, Polyclonal Antibody, Abnova™

Product Code. 16263290
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16263290 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16263290 Supplier Abnova Supplier No. PAB13487.100uL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit polyclonal antibody raised against native Col1a1/Col1a2.

Specifications

Antigen Col1a1/Col1a2
Applications ELISA, Immunofluorescence, Immunohistochemistry (PFA fixed), Radioimmune Assays (RIA)
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against native Col1a1/Col1a2.
Dilution Immunohistochemistry (1:200-1:600) Immunofluorescence (1:40) The optimal working dilution should be determined by the end user.
Formulation In phosphate buffer
Gene Alias COLIA1
Gene Symbols Col1a1
Host Species Rabbit
Immunogen Native purified Col1a1/Col1a2 from rat skin.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29393/84352
Test Specificity This antibody reacts with rat collagen type I 100%, rat collagen II, III, V: <0.1%; human, mouse, chicken collagen type I <0.1%; rat elastin <0.1% ( RIA at 1:200 dilution).
Target Species Rat
Content And Storage Store at -20°C.
After reconstitution with deionized water, store at -20°C or lower.
Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.