missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complement C4c, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against native Complement C4c.
Brand: Abnova PAB19886.150ug
This item is not returnable.
View return policy
Specifications
| Complement C4c | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against native Complement C4c. | |
| In PBS, pH 7.5 (0.02% sodium azide, 50% glycerol (v/v)) | |
| Native purified complement C4c from human plasma. | |
| 150 μg | |
| Primary | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| Rabbit | |
| Affinity purification | |
| RUO | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction