Learn More
COPB2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant COPB2.
Brand: Abnova PAB23074.100uL
Description
The Golgi coatomer complex (see MIM 601924) constitutes the coat of nonclathrin-coated vesicles and is essential for Golgi budding and vesicular trafficking. It consists of 7 protein subunits, including COPB2.[supplied by OMIM
Sequence: VALGYDEGSIIVKLGREEPAMSMDANGKIIWAKHSEVQQANLKAMGDAEIKDGERLPLAVKDMGSCEIYPQTIQHNPNGRFVVVCGDGEYIIYTASpecifications
| COPB2 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against recombinant COPB2. | |
| In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
| P35606 | |
| COPB2 | |
| Recombinant protein corresponding to amino acids of human COPB2. | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry, Western Blot, Western Blot | |
| Unconjugated | |
| Immunohistochemistry (1:50-1:200) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user. | |
| COPB2 | |
| beta'-COP | |
| Rabbit | |
| Antigen affinity purification | |
| RUO | |
| 9276 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only