Learn More
CREB1 (phospho S133), Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic phosphopeptide of CREB1.
Brand: Abnova PAB12617.100ug
Description
This gene encodes a transcription factor that is a member of the leucine zipper family of DNA binding proteins. This protein binds as a homodimer to the cAMP-responsive element, an octameric palindrome. The protein is phosphorylated by several protein kinases, and induces transcription of genes in response to hormonal stimulation of the cAMP pathway. Alternate splicing of this gene results in two transcript variants encoding different isoforms. [provided by RefSeq
Sequence: RPSYRSpecifications
| CREB1 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic phosphopeptide of CREB1. | |
| In TBS, pH 7.2 (BSA, 10% Proclin300) | |
| CREB/MGC9284 | |
| Rabbit | |
| 100 μg | |
| Primary | |
| This antibody only recognizes ∼40 KDa of human CREB1. | |
| Store at 4°C. For long term storage store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| Dot Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Western Blot (0.1-1 ug/mL) ELISA (0.01-0.1 ug/mL) Immunoprecipitation (2-5 ug/mL) Immunohistochemistry (1:50) The optimal working dilution should be determined by the end user. | |
| CREB1 | |
| CREB1 | |
| Synthetic phosphopeptide corresponding to residues surrounding S133 of human CREB1. | |
| RUO | |
| 1385 | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only