missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CREB3L4, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of CREB3L4.
Brand: Abnova PAB19819.100ug
This item is not returnable.
View return policy
Description
cAMP response element-binding (CREB) proteins are a family of mammalian transcription activators. For further background information on CREB proteins, see CREB1 (MIM 123810).[supplied by OMIM
Sequence: PSGRIRSVLHADEMSpecifications
| CREB3L4 | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against synthetic peptide of CREB3L4. | |
| In serum (0.02% sodium azide) | |
| Q8TEY5 | |
| CREB3L4 | |
| A synthetic peptide corresponding to C-terminus of human CREB3L4. | |
| RUO | |
| 148327 | |
| Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| ELISA (1:80000) Western Blot (1:200-1000) The optimal working dilution should be determined by the end user. | |
| CREB3L4 | |
| AIBZIP/ATCE1/CREB3/CREB4/JAL/hJAL | |
| Rabbit | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction