missing translation for 'onlineSavingsMsg'
Learn More

CSF3 Rabbit anti-Human, Polyclonal Antibody, Abnova™

Product Code. 16293810
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16293810 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16293810 Supplier Abnova Supplier No. PAB19561.100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit polyclonal antibody raised against recombinant CSF3.

The protein encoded by this gene is a cytokine that controls the production, differentiation, and function of granulocytes. The active protein is found extracellularly. Three transcript variants encoding three different isoforms have been found for this gene. [provided by RefSeq

Specifications

Antigen CSF3
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against recombinant CSF3.
Dilution ELISA (0.5-1 ug/mL; The detection limit is 0.00245 ng/well) Western Blot (1-2 ug/mL) Immunohistochemistry (2-10 ug/mL) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from PBS.
Gene CSF3
Gene Accession No. P09919
Gene Alias G-CSF/GCSF/MGC45931
Gene Symbols CSF3
Host Species Rabbit
Immunogen Recombinant protein corresponding to human CSF3.
Purification Method Protein G purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1440
Target Species Human
Content And Storage Store at -20°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.